3-Ethynyl-1-tosylazetidin-3-ol
Lieferant: Apollo Scientific
OR312054-1G
APOSOR312054-1GEA
519
EUR
APOSOR312054-1G
3-Ethynyl-1-tosylazetidin-3-ol
3-Ethynyl-1-tosylazetidin-3-ol
Formel:
C₁₂H₁₃NO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD22421707 CAS-Nummer: 1349199-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...