Ethyl 2-(thietan-3-ylidene)acetate

Lieferant: Apollo Scientific
Achtung

OR312060-1G
APOSOR312060-1GEA 306 EUR
APOSOR312060-1G
Ethyl 2-(thietan-3-ylidene)acetate
Ethyl 2-(thietan-3-ylidene)acetate
Formel: C₇H₁₀O₂S
Molecular Weight: 158,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12755198
CAS-Nummer: 1223573-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR