1-(Oxetan-3-yl)piperazine

Lieferant: Apollo Scientific
Achtung

OR312090-1G
APOSOR312090-1GEA 77 EUR
APOSOR312090-1G
1-(Oxetan-3-yl)piperazine
1-(Oxetan-3-yl)piperazine
Formel: C₇H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD21847918
CAS-Nummer: 1254115-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR