4-(3-Azetidinyl)morpholine dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR312091-1G
APOSOR312091-1GEA 0 EUR
APOSOR312091-1G
4-(3-Azetidinyl)morpholine dihydrochloride
4-(3-Azetidinyl)morpholine dihydrochloride
Formel: C₇H₁₆Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11617823
CAS-Nummer: 178312-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR