(1-Aminocyclobut-1-yl)acetic acid, N-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1-((tert-Butoxycarbonyl)amino)cyclobutyl)acetic acid, 2-(1-Aminocyclobut-1-yl)ethanoic acid, N-BOC protected, {1-[(tert-Butoxycarbonyl)amino]cyclobut-1-yl}acetic acid
OR312103-250MG
APOSOR312103-250MGEA
77
EUR
APOSOR312103-250MG
(1-Aminocyclobut-1-yl)acetic acid, N-BOC protected
(1-Aminocyclobut-1-yl)acetic acid, N-BOC protected
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229,273 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD13249390 CAS-Nummer: 249762-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...