Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR312111-1G OR312111-5G
APOSOR312111-1GEA 71.2 EUR
APOSOR312111-1G APOSOR312111-5G
Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 98%
Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid
Formel: C₇H₈O₄
Molecular Weight: 156.136 g/mol
Schmelzpunkt: 302…305 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD20621127
CAS-Nummer: 56842-95-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR