2-[1-(Aminomethyl)cyclobutyl]acetic acid hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR312117-100MG
APOSOR312117-100MGEA 278 EUR
APOSOR312117-100MG
2-[1-(Aminomethyl)cyclobutyl]acetic acid hydrochloride
2-[1-(Aminomethyl)cyclobutyl]acetic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20645823
CAS-Nummer: 748754-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR