3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane 97%

Lieferant: Apollo Scientific

Synonyme: (3-(Pyrrolidin-1-yl)oxetan-3-yl)methanamine

OR312143-250MG
APOSOR312143-250MGEA 511 EUR
APOSOR312143-250MG
3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane 97%
3-Aminomethyl-3-(pyrrolidin-1-yl)oxetane
Formel: C₈H₁₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD22628718
CAS-Nummer: 1416323-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR