trans-3-(Boc-amino)cyclobutanecarboxylic acid 98%

Lieferant: Apollo Scientific

OR312151-1G OR312151-5G
APOSOR312151-1GEA 0 EUR
APOSOR312151-1G APOSOR312151-5G
trans-3-(Boc-amino)cyclobutanecarboxylic acid 98%
trans-3-(Boc-amino)cyclobutanecarboxylic acid
Formel: C₁₀H₁₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06657680
CAS-Nummer: 939400-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR