N-Phenyloxetan-3-amine
Lieferant: Apollo Scientific
OR312180-1G
APOSOR312180-1GEA
2030
EUR
APOSOR312180-1G
N-Phenyloxetan-3-amine
N-Phenyloxetan-3-amine
Formel:
C₉H₁₁NO Molecular Weight: 149,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18849128 CAS-Nummer: 1341385-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...