1,1-Cyclobutanedimethanol 95%

Lieferant: Apollo Scientific
Gefahr

OR312194-1G
APOSOR312194-1GEA 0 EUR
APOSOR312194-1G
1,1-Cyclobutanedimethanol 95%
1,1-Cyclobutanedimethanol
Formel: C₆H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00463706
CAS-Nummer: 4415-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR