1,4,8,11-Tetraazacyclotetradecan 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
CYCLAM
OR3122-1G
APOSOR3122-1GEA
23.2
EUR
APOSOR3122-1G
1,4,8,11-Tetraazacyclotetradecan 98%
1,4,8,11-Tetraazacyclotetradecane
Formel:
C₁₀H₂₄N₄ Molecular Weight: 200,33 g/mol Schmelzpunkt: 184…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005105 CAS-Nummer: 295-37-4 EINECS: 206-039-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...