tert-Butyl 2,6-diazaspiro[3.5]nonane-6-carboxylate

Lieferant: Apollo Scientific

Synonyme: 2,6-Diazaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester

OR312205-250MG
APOSOR312205-250MGEA 275 EUR
APOSOR312205-250MG
tert-Butyl 2,6-diazaspiro[3.5]nonane-6-carboxylate
tert-Butyl 2,6-diazaspiro[3.5]nonane-6-carboxylate
Formel: C₁₂H₂₂N₂O₂
Molecular Weight: 226,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234981
CAS-Nummer: 885272-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR