2-Oxa-6-azaspiro[3.4]octan-7-one

Lieferant: Apollo Scientific

OR312208-250MG
APOSOR312208-250MGEA 194 EUR
APOSOR312208-250MG
2-Oxa-6-azaspiro[3.4]octan-7-one
2-Oxa-6-azaspiro[3.4]octan-7-one
Formel: C₆H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14586414
CAS-Nummer: 1207174-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR