1,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 1,7-diazaspiro[3.5]nonane-7-carboxylate

OR312211-250MG
APOSOR312211-250MGEA 161 EUR
APOSOR312211-250MG
1,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester
1,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester
Formel: C₁₂H₂₂N₂O₂
Molecular Weight: 226,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14581195
CAS-Nummer: 1180112-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR