2,5-Diazaspiro[3.4]octane-5-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2,5-diazaspiro[3.4]octane-5-carboxylate

OR312236-250MG
APOSOR312236-250MGEA 490 EUR
APOSOR312236-250MG
2,5-Diazaspiro[3.4]octane-5-carboxylic acid tert-butyl ester
2,5-Diazaspiro[3.4]octane-5-carboxylic acid tert-butyl ester
Formel: C₁₁H₂₀N₂O₂
Molecular Weight: 212,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501186
CAS-Nummer: 1086398-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR