6-Azaspiro[2.5]octane hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-azaspiro[2.5]octane hydrochloride

OR312258-5G
APOSOR312258-5GEA 111 EUR
APOSOR312258-5G
6-Azaspiro[2.5]octane hydrochloride
6-Azaspiro[2.5]octane hydrochloride
Formel: C₇H₁₃N·ClH
Molecular Weight: 147.646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14581272
CAS-Nummer: 1037834-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR