7-Azaspiro[3.5]nonane

Lieferant: Apollo Scientific
Achtung

OR312259-250MG
APOSOR312259-250MGEA 783 EUR
APOSOR312259-250MG
7-Azaspiro[3.5]nonane
7-Azaspiro[3.5]nonane
Formel: C₈H₁₅N
Molecular Weight: 125,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14581275
CAS-Nummer: 766-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR