2-Oxa-spiro[3.3]heptan-6-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxaspiro[3.3]heptan-6-one

OR312273-1G
APOSOR312273-1GEA 899 EUR
APOSOR312273-1G
2-Oxa-spiro[3.3]heptan-6-one
2-Oxa-spiro[3.3]heptan-6-one
Formel: C₆H₈O₂
Molecular Weight: 112,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22415239
CAS-Nummer: 1339892-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR