5-Oxa-2-azaspiro[3.5]nonane oxalate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Oxa-2-azaspiro[3.5]nonane ethane-1,2-dioate
OR312275-100MG
APOSOR312275-100MGEA
65.4
EUR
APOSOR312275-100MG
5-Oxa-2-azaspiro[3.5]nonane oxalate 95%
5-Oxa-2-azaspiro[3.5]nonane oxalate
Formel:
C₇H₁₃NO·C₂H₂O₄ Molecular Weight: 217,219 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20926175 CAS-Nummer: 1427359-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...