5-Methyl-2-oxa-5,8-diazaspiro[3.5]nonane

Lieferant: Apollo Scientific
Achtung

OR312278-100MG
APOSOR312278-100MGEA 1210 EUR
APOSOR312278-100MG
5-Methyl-2-oxa-5,8-diazaspiro[3.5]nonane
5-Methyl-2-oxa-5,8-diazaspiro[3.5]nonane
Formel: C₇H₁₄N₂O
Molecular Weight: 142,199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22070771
CAS-Nummer: 1369103-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR