Benzyl 2,6-diazaspiro[3.4]octane-2-carboxylate 95+%

Lieferant: Apollo Scientific

OR312300-250MG
APOSOR312300-250MGEA 151 EUR
APOSOR312300-250MG
Benzyl 2,6-diazaspiro[3.4]octane-2-carboxylate 95+%
Benzyl 2,6-diazaspiro[3.4]octane-2-carboxylate
Formel: C₁₄H₁₈N₂O₂
Molecular Weight: 246,305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223550
CAS-Nummer: 1086394-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR