tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Boc-1,8-diaza-spiro[4.5]decane

OR312325-250MG
APOSOR312325-250MGEA 140 EUR
APOSOR312325-250MG
tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate 95+%
tert-Butyl 1,8-diazaspiro[4.5]decane-1-carboxylate
Formel: C₁₃H₂₄N₂O₂
Molecular Weight: 240,342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739037
CAS-Nummer: 885279-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR