2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2,9-diazaspiro[5.5]undecane-2-carboxylate

OR312332-100MG
APOSOR312332-100MGEA 173 EUR
APOSOR312332-100MG
2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester
2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester
Formel: C₁₄H₂₆N₂O₂
Molecular Weight: 254,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223510
CAS-Nummer: 189333-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR