Methyl 3-(2-amino-1,3-oxazol-4-yl)bicyclo[1.1.1]pentane-1-carboxylate

Lieferant: Apollo Scientific

OR312427-250MG
APOSOR312427-250MGEA 1940 EUR
APOSOR312427-250MG
Methyl 3-(2-amino-1,3-oxazol-4-yl)bicyclo[1.1.1]pentane-1-carboxylate
Methyl 3-(2-amino-1,3-oxazol-4-yl)bicyclo[1.1.1]pentane-1-carboxylate
Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037318
CAS-Nummer: 1980063-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR