3-(tert-Butoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid

Lieferant: Apollo Scientific

OR312440-250MG
APOSOR312440-250MGEA 271 EUR
APOSOR312440-250MG
3-(tert-Butoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
3-(tert-Butoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₁₁H₁₆O₄
Molecular Weight: 212.242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27987015
CAS-Nummer: 1638765-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR