3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid

Lieferant: Apollo Scientific

OR312451-250MG
APOSOR312451-250MGEA 502 EUR
APOSOR312451-250MG
3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid
3-Methylbicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₇H₁₀O₂
Molecular Weight: 126.153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19228632
CAS-Nummer: 65862-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR