Bicyclo[1.1.1]pentane-1-carbonitrile

Lieferant: Apollo Scientific
Gefahr

OR312471-1G
APOSOR312471-1GEA 4730 EUR
APOSOR312471-1G
Bicyclo[1.1.1]pentane-1-carbonitrile
Bicyclo[1.1.1]pentane-1-carbonitrile
Formel: C₆H₇N
Molecular Weight: 93,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037333
CAS-Nummer: 126332-44-3
UN: 1992
ADR: 3+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR