3-Acetylbicyclo[1.1.1]pentane-1-carboxylic acid

Lieferant: Apollo Scientific

OR312477-250MG
APOSOR312477-250MGEA 1080 EUR
APOSOR312477-250MG
3-Acetylbicyclo[1.1.1]pentane-1-carboxylic acid
3-Acetylbicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₈H₁₀O₃
Molecular Weight: 154.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18826733
CAS-Nummer: 156329-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR