4-(Methoxycarbonyl)cubanecarboxylic acid
Lieferant: Apollo Scientific
OR312489-250MG
APOSOR312489-250MGEA
0
EUR
APOSOR312489-250MG
4-(Methoxycarbonyl)cubanecarboxylic acid
4-(Methoxycarbonyl)cubanecarboxylic acid
Formel:
C₁₁H₁₀O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458189 CAS-Nummer: 24539-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...