Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-aminobicyclo[2.2.2]octane-1-carboxylate hydrochloride, 4-Aminobicyclo[2.2.2]octane-1-carboxylic acid methyl ester

OR312511-250MG
APOSOR312511-250MGEA 75.4 EUR
APOSOR312511-250MG
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate
Methyl-4-aminobicyclo[2.2.2]octane-1-carboxylate
Formel: C₁₀H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20693756
CAS-Nummer: 135908-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR