4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carbonsäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Bicyclo[2.2.2]octane-1,4-dicarboxylic acid monomethyl ester, Monomethyl Hydrogen Bicyclo[2.2.2]octane-1,4-dicarboxylate, 4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carboxylic acid
OR312513-1G
APOSOR312513-1GEA
21.9
EUR
APOSOR312513-1G
4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carbonsäure
4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid
Formel:
C₁₁H₁₆O₄ Molecular Weight: 212,2423 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18459359 CAS-Nummer: 18720-35-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...