Trimorpholinophosphine oxide 97%

Lieferant: Apollo Scientific

OR3128-100G OR3128-25G
APOSOR3128-100GEA 0 EUR
APOSOR3128-100G APOSOR3128-25G
Trimorpholinophosphine oxide 97%
Trimorpholinophosphine oxide
Formel: C₁₂H₂₄N₃O₄P
Lagertemperatur: Raumtemperatur
MDL: MFCD00047406
CAS-Nummer: 4441-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR