(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected

Lieferant: Apollo Scientific
Achtung

OR313003-1G OR313003-250MG
APOSOR313003-1GEA 0 EUR
APOSOR313003-1G APOSOR313003-250MG
(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected
(3R)-3-(Bromomethyl)pyrrolidine, N-BOC protected
Formel: C₁₀H₁₈BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08059335
CAS-Nummer: 1067230-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR