Spiro[isoindole-1,4'-piperidin]-3(2H)-one hydrochloride

Lieferant: Apollo Scientific
Achtung

OR313019-1G
APOSOR313019-1GEA 1360 EUR
APOSOR313019-1G
Spiro[isoindole-1,4'-piperidin]-3(2H)-one hydrochloride
Spiro[isoindole-1,4'-piperidin]-3(2H)-one hydrochloride
Formel: C₁₂H₁₄N₂O·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD18711590
CAS-Nummer: 788812-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR