1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

OR313025-1G
APOSOR313025-1GEA 55.1 EUR
APOSOR313025-1G
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1-Cyclopropyl-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₂H₁₉BN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD16659007
CAS-Nummer: 1151802-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR