2-(4-Bromophenyl)pyrrolidine 97%

Lieferant: Apollo Scientific
Achtung

OR313026-1G
APOSOR313026-1GEA 0 EUR
APOSOR313026-1G
2-(4-Bromophenyl)pyrrolidine 97%
2-(4-Bromophenyl)pyrrolidine
Formel: C₁₀H₁₂BrN
Siedepunkt: 287,2 °C/760mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD02663432
CAS-Nummer: 383127-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR