N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid

OR313034-1G OR313034-250MG
APOSOR313034-1GEA 289 EUR
APOSOR313034-1G APOSOR313034-250MG
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
Formel: C₁₅H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD05861548
CAS-Nummer: 170097-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR