N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
OR313034-1G
OR313034-250MG
APOSOR313034-1GEA
289
EUR
APOSOR313034-1G
APOSOR313034-250MG
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
N-Boc-1,2,3,4-tetrahydroisoquinoline-6-carboxylic acid
Formel:
C₁₅H₁₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861548 CAS-Nummer: 170097-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...