2-Methyl-2-propanyl-3-formyl-1-pyrrolidincarboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (+/-)-3-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, (+/-)-1-Boc-pyrrolidine-3-carboxaldehyde, (+/-)-1-(tert-Butoxycarbonyl)-3-formylpyrrolidine, (+/-)-1-Boc-3-formylpyrrolidine, tert-Butyl 3-formylpyrrolidine-1-carboxylate, (+/-)-tert-Butyl 3-Formylpyrrolidine-1-carboxylate, (+/-)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxaldehyde

OR313035-5G
APOSOR313035-5GEA 0 EUR
APOSOR313035-5G
2-Methyl-2-propanyl-3-formyl-1-pyrrolidincarboxylat 98%
tert-Butyl-3-formylpyrrolidine-1-carboxylate
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199.2500 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03791257
CAS-Nummer: 59379-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR