Ethyl-3-methyl-1H-pyrrole-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-methyl-1H-pyrrole-2-carboxylate

OR313039-1G OR313039-5G OR313039-25G
APOSOR313039-1GEA 77 EUR
APOSOR313039-1G APOSOR313039-5G APOSOR313039-25G
Ethyl-3-methyl-1H-pyrrole-2-carboxylate 98%
Ethyl-3-methyl-1H-pyrrole-2-carboxylate
Formel: C₈H₁₁NO₂
Molecular Weight: 153.1784 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01006747
CAS-Nummer: 3284-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR