1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol

Lieferant: Apollo Scientific
Achtung

OR313043-1G
APOSOR313043-1GEA 136 EUR
APOSOR313043-1G
1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol
1-(3,4-Dimethoxyphenyl)-2-(2-methoxyphenoxy)propane-1,3-diol
Formel: C₁₈H₂₂O₆
Molecular Weight: 334,3600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01880862
CAS-Nummer: 10535-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR