2-Formyl-5-methoxypyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methoxypyridine-2-carbaldehyde, 5-Methoxypicolinaldehyde

OR313046-1G
APOSOR313046-1GEA 75.4 EUR
APOSOR313046-1G
2-Formyl-5-methoxypyridin
2-Formyl-5-methoxypyridin
Formel: C₇H₇NO₂
Molecular Weight: 137.1360 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234867
CAS-Nummer: 22187-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR