N-(2-Aminoethyl)maleimide hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2-Aminoethyl)maleimide HCl, N-(2-Aminoethyl)maleimide Hydrochloride, 2-Maleimidoethylamine hydrochloride

OR313050-5G
APOSOR313050-5GEA 127 EUR
APOSOR313050-5G
N-(2-Aminoethyl)maleimide hydrochloride
N-(2-Aminoethyl)maleimide hydrochloride
Formel: C₆H₈N₂O₂
Molecular Weight: 176.60 g/mol
Schmelzpunkt: 163 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD09800491
CAS-Nummer: 134272-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR