(R)-2-Methylpiperidine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR313055-1G
APOSOR313055-1GEA
183
EUR
APOSOR313055-1G
(R)-2-Methylpiperidine 97%
(R)-2-Methylpiperidine
Formel:
C₆H₁₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862168 CAS-Nummer: 1722-95-8 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...