5-Nitro-2,4,6-trichloropyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Nitro-2,4,6-trichloro-1,3-diazine

OR313061-5G
APOSOR313061-5GEA 209 EUR
APOSOR313061-5G
5-Nitro-2,4,6-trichloropyrimidine
5-Nitro-2,4,6-trichloropyrimidine
Formel: C₄Cl₃N₃O₂
Molecular Weight: 228.421 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00091516
CAS-Nummer: 4359-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR