3-Cyclopropylpropionic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR313064-250MG
APOSOR313064-250MGEA
65.4
EUR
APOSOR313064-250MG
3-Cyclopropylpropionic acid 95%
3-Cyclopropylpropionic acid
Formel:
C₆H₁₀O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD02179408 CAS-Nummer: 5618-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...