(S)-3-Methyl-pyrrolidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR313067-1G
OR313067-250MG
APOSOR313067-1GEA
0
EUR
APOSOR313067-1G
APOSOR313067-250MG
(S)-3-Methyl-pyrrolidine hydrochloride
(S)-3-Methyl-pyrrolidine hydrochloride
Formel:
C₅H₁₁N·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796632 CAS-Nummer: 186597-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...