(Pyrazin-2-yl)acetic acid 97%

Lieferant: Apollo Scientific
Achtung

OR313069-1G OR313069-5G
APOSOR313069-1GEA 0 EUR
APOSOR313069-1G APOSOR313069-5G
(Pyrazin-2-yl)acetic acid 97%
(Pyrazin-2-yl)acetic acid
Formel: C₆H₆N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD06796638
CAS-Nummer: 140914-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR