Methyl 5-bromo-6-methoxynicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-bromo-6-methoxy-3-pyridinecarboxylic acid methyl ester

OR313072-5G
APOSOR313072-5GEA 155 EUR
APOSOR313072-5G
Methyl 5-bromo-6-methoxynicotinate
Methyl 5-bromo-6-methoxynicotinate
Formel: C₈H₈BrNO₃
Molecular Weight: 246,058 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14698128
CAS-Nummer: 93349-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR